Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-methoxy-4-(trifluoromethoxy)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

32325-500MG 32325-1G 32325-5G
77339-472EA 593.28 USD InStock
77339-472 77349-078 77373-334
Ethyl 3-methoxy-4-(trifluoromethoxy)benzoate ≥97%
Ethyl 3-methoxy-4-(trifluoromethoxy)benzoate


Formula: C₁₁H₁₁F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD18396708
CAS Number: 1261582-69-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty