Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium 2-iodophenyltrifluoroborate ≥97%

Supplier: AOB CHEM USA
Warning

33028-500MG 33028-1G 33028-5G 33028-25G
77325-448EA 92.7 USD InStock
77325-448 77342-460 77361-812 77373-426
Potassium 2-iodophenyltrifluoroborate ≥97%
Potassium 2-iodophenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₆H₄BF₃IK
Storage Temperature: Ambient
MDL Number: MFCD30723166
CAS Number: 1431616-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty