Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-isopropoxybenzoic acid methyl ester ≥97%

Supplier: AOB CHEM USA
Warning

33402-500MG 33402-1G 33402-5G
77336-340EA 387.28 USD InStock
77336-340 77349-212 77373-462
2-Fluoro-4-isopropoxybenzoic acid methyl ester ≥97%
2-Fluoro-4-isopropoxybenzoic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD23115008
CAS Number: 1394933-49-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty