Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propan-2-yl 3,5-dibromopyridine-4-carboxylate ≥97%

Supplier: AOB CHEM USA
Warning

33565-500MG 33565-1G 33565-5G 33565-25G
77325-532EA 201.88 USD InStock
77325-532 77349-224 77361-902 77384-958
Propan-2-yl 3,5-dibromopyridine-4-carboxylate ≥97%
Propan-2-yl 3,5-dibromopyridine-4-carboxylate
Precaución: for Research Use Only


Formula: C₉H₉Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD30834080
CAS Number: 2121512-81-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty