Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenylmethyl 3,5-dibromopyridine-4-carboxylate ≥97%

Supplier: AOB CHEM USA
Warning

33566-500MG 33566-1G 33566-5G 33566-25G
77325-534EA 210.12 USD InStock
77325-534 77349-226 77361-904 77384-960
Phenylmethyl 3,5-dibromopyridine-4-carboxylate ≥97%
Phenylmethyl 3,5-dibromopyridine-4-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₉Br₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD30834081
CAS Number: 2121512-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty