Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-fluoro-3-methylthiophenylboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

33760-500MG 33760-1G 33760-5G 33760-25G
77325-574EA 91.67 USD InStock
77325-574 77342-420 77361-948 77373-508
4-Chloro-2-fluoro-3-methylthiophenylboronic acid ≥97%
4-Chloro-2-fluoro-3-methylthiophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BClFO₂S
Storage Temperature: Ambient
MDL Number: MFCD22573253
CAS Number: 944128-91-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty