Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzylcarbamoyl-2-chlorobenzeneboronic acid ≥97%

Supplier: AOB CHEM USA
Warning

34090-500MG 34090-1G 34090-25G 34090-5G
77325-626EA 297.67 USD InStock
77325-626 77349-304 77385-022 77373-556
4-Benzylcarbamoyl-2-chlorobenzeneboronic acid ≥97%
4-Benzylcarbamoyl-2-chlorophenylboronic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₃BClNO₃
MW: 289.522 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22125049
CAS Number: 2096334-06-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty