Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenoxathiin-4-boronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

35243-1G 35243-5G 35243-25G
77341-082EA 70.04 USD InStock
77341-082 77362-140 77373-706
Phenoxathiin-4-boronic acid pinacol ester ≥97%
Phenoxathiin-4-boronic acid pinacol ester


Formula: C₁₈H₁₉BO₃S
Storage Temperature: Ambient
MDL Number: MFCD32707006
CAS Number: 2290561-93-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty