Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-(2-methoxyethoxy)nicotinate ≥97%

Supplier: AOB CHEM USA
Warning

36860-500MG 36860-1G 36860-5G 36860-25G
77326-224EA 245.14 USD InStock
77326-224 77349-922 77362-550 77385-434
Methyl 6-(2-methoxyethoxy)nicotinate ≥97%
Methyl 6-(2-methoxyethoxy)nicotinate
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₄
MW: 211.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632091
CAS Number: 2379322-08-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty