Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-(benzyloxy)-3,5-dichlorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

37544-500MG 37544-1G 37544-5G 37544-25G
77326-424EA 93.73 USD InStock
77326-424 77342-502 77362-738 77374-316
Ethyl 4-(benzyloxy)-3,5-dichlorobenzoate ≥97%
Ethyl 4-(benzyloxy)-3,5-dichlorobenzoate
Precaución: for Research Use Only


Formula: C₁₆H₁₄Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD06204971
CAS Number: 40689-39-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty