Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(benzyloxy)-3,5-dichlorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

37545-500MG 37545-1G 37545-5G 37545-25G
77326-426EA 87.55 USD InStock
77326-426 77342-290 77362-740 77374-318
Methyl 4-(benzyloxy)-3,5-dichlorobenzoate ≥97%
Methyl 4-(benzyloxy)-3,5-dichlorobenzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₂Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD06204057
CAS Number: 536974-83-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty