Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,5-difluoro-4-propoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

37862-500MG 37862-1G 37862-5G
77338-594EA 503.67 USD InStock
77338-594 77350-182 77374-386
Methyl 2,5-difluoro-4-propoxybenzoate ≥97%
Methyl 2,5-difluoro-4-propoxybenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₂F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34564053
CAS Number: 2734774-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty