Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3-bromo-5-chlorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

37948-500MG 37948-1G 37948-5G 37948-25G
77326-534EA 181.28 USD InStock
77326-534 77346-832 77362-826 77385-648
tert-Butyl 3-bromo-5-chlorobenzoate ≥97%
tert-Butyl 3-bromo-5-chlorobenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₂BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD33023827
CAS Number: 1870857-76-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty