Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Fluoro-6-methoxypyridin-3-yl)boronic acid ≥97%

Supplier: AOB CHEM USA
Warning

41969-500MG 41969-1G 41969-5G 41969-25G
77326-894EA 91.67 USD InStock
77326-894 77342-432 77363-188 77374-726
(2-Fluoro-6-methoxypyridin-3-yl)boronic acid ≥97%
(2-Fluoro-6-methoxypyridin-3-yl)boronic acid
Precaución: for Research Use Only


Formula: C₆H₇BFNO₃
MW: 170.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611244
CAS Number: 1402238-30-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty