Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Bromo-2-chloro-5-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%

Supplier: AOB CHEM USA
Warning

45471-500MG 45471-1G 45471-5G 45471-25G
77326-936EA 277.07 USD InStock
77326-936 77350-558 77363-228 77385-932
2-(4-Bromo-2-chloro-5-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%
2-(4-Bromo-2-chloro-5-(trifluoromethyl)phenyl)-1,3-dioxolane
Precaución: for Research Use Only


Formula: C₁₀H₇BrClF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD32707118
CAS Number: 2432849-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty