Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(benzyloxy)-3-bromo-5-methylbenzoate ≥95%

Supplier: AOB CHEM USA
Warning

45922-500MG 45922-1G 45922-5G 45922-25G
77327-004EA 243.08 USD InStock
77327-004 77350-614 77363-306 77385-976
Methyl 2-(benzyloxy)-3-bromo-5-methylbenzoate ≥95%
Methyl 2-(benzyloxy)-3-bromo-5-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₆H₁₅BrO₃
MW: 335.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707142
CAS Number: 2432848-62-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty