Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

46194-500MG 46194-1G 46194-5G 46194-25G
77327-096EA 263.68 USD InStock
77327-096 77350-672 77363-390 77386-030
3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%
3,5-Difluoro-4'-methyl-[1,1'-biphenyl]-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₀F₂O₂
MW: 248.225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902697
CAS Number: 2361379-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty