Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-(benzyloxy)-3-methylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

47513-500MG 47513-1G 47513-5G 47513-25G
77327-382EA 281.19 USD InStock
77327-382 77350-946 77363-726 77386-214
Ethyl 4-(benzyloxy)-3-methylbenzoate ≥97%
Ethyl 4-(benzyloxy)-3-methylbenzoate
Precaución: for Research Use Only


Formula: C₁₇H₁₈O₃
MW: 270.323 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403858
CAS Number: 158771-02-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty