Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethyl(naphthalen-1-yl)silane ≥97%

Supplier: AOB CHEM USA
Warning

47644-500MG 47644-1G 47644-5G 47644-25G
77327-398EA 220.42 USD InStock
77327-398 77350-964 77363-742 77386-224
Trimethyl(naphthalen-1-yl)silane ≥97%
Trimethyl(naphthalen-1-yl)silane
Precaución: for Research Use Only


Formula: C₁₃H₁₆Si
MW: 200.352 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00094131
CAS Number: 18052-80-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty