Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3,5-dichloro-4-propoxybenzoate ≥95%

Supplier: AOB CHEM USA
Warning

47950-500MG 47950-1G 47950-5G 47950-25G
77327-412EA 114.33 USD InStock
77327-412 77343-356 77363-764 77375-326
Ethyl 3,5-dichloro-4-propoxybenzoate ≥95%
Ethyl 3,5-dichloro-4-propoxybenzoate
Precaución: for Research Use Only


Formula: C₁₂H₁₄Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD01765356
CAS Number: 40689-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty