Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-chloro-3-ethoxy-2-fluorobenzoate ≥97%

Supplier: AOB CHEM USA
Warning

48426-500MG 48426-1G 48426-5G 48426-25G
77327-566EA 278.1 USD InStock
77327-566 77351-162 77363-906 77386-342
Methyl 6-chloro-3-ethoxy-2-fluorobenzoate ≥97%
Methyl 6-chloro-3-ethoxy-2-fluorobenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD33403883
CAS Number: 2385337-05-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty