Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-(methoxymethyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

48499-500MG 48499-1G 48499-5G 48499-25G
77327-582EA 261.62 USD InStock
77327-582 77351-178 77363-920 77386-358
4-Fluoro-3-(methoxymethyl)benzoic acid ≥97%
4-Fluoro-3-(methoxymethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD17287385
CAS Number: 1333834-34-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty