Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-bromo-2,5-dichlorobenzoate ≥95%

Supplier: AOB CHEM USA
Warning

48972-500MG 48972-1G 48972-5G 48972-25G
77327-808EA 100.94 USD InStock
77327-808 77342-922 77364-130 77375-624
Ethyl 4-bromo-2,5-dichlorobenzoate ≥95%
Ethyl 4-bromo-2,5-dichlorobenzoate
Precaución: for Research Use Only


Formula: C₉H₇BrCl₂O₂
MW: 297.961 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24108584
CAS Number: 1538275-43-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty