Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(benzyloxy)-2,6-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOB CHEM USA
Warning

48991-500MG 48991-1G 48991-5G 48991-25G
77327-814EA 141.11 USD InStock
77327-814 77344-808 77364-138 77386-528
2-(4-(benzyloxy)-2,6-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-(benzyloxy)-2,6-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₂₁H₂₇BO₃
MW: 338.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178559
CAS Number: 1823170-16-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty