Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Fluorobenzoyl)benzaldehyde 97

Supplier: AOB CHEM USA
Warning

49086-500MG 49086-1G 49086-5G 49086-25G
77327-850EA 119.48 USD InStock
77327-850 77343-648 77364-168 77375-650
3-(4-Fluorobenzoyl)benzaldehyde 97
3-(4-Fluorobenzoyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₄H₉FO₂
MW: 228.218 g/mol
Storage Temperature: Ambient
CAS Number: 75894-14-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty