Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3,4-difluoro-2-methoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

49134-500MG 49134-1G 49134-5G 49134-25G
77327-872EA 138.02 USD InStock
77327-872 77344-650 77364-192 77386-572
Ethyl 3,4-difluoro-2-methoxybenzoate ≥97%
Ethyl 3,4-difluoro-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11520401
CAS Number: 1803847-67-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty