Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluoro-3-methoxy-N,N-dimethylbenzamide ≥97%

Supplier: AOB CHEM USA
Warning

49430-500MG 49430-1G 49430-5G 49430-25G
77327-964EA 189.52 USD InStock
77327-964 77351-486 77364-282 77386-630
5-Bromo-2-fluoro-3-methoxy-N,N-dimethylbenzamide ≥97%
5-Bromo-2-fluoro-3-methoxy-N,N-dimethylbenzamide
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrFNO₂
MW: 276.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902827
CAS Number: 2586125-87-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty