Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,6-dichloro-3-isopropoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

51252-500MG 51252-1G 51252-5G 51252-25G
77328-322EA 278.1 USD InStock
77328-322 77351-882 77364-658 77386-884
Methyl 2,6-dichloro-3-isopropoxybenzoate ≥97%
Methyl 2,6-dichloro-3-isopropoxybenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₂Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34530074
CAS Number: 2734774-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty