Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-fluoro-3-methoxy-6-(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

51374-500MG 51374-1G 51374-5G 51374-25G
77328-366EA 141.11 USD InStock
77328-366 77344-820 77364-708 77386-900
Ethyl 2-fluoro-3-methoxy-6-(trifluoromethyl)benzoate ≥97%
Ethyl 2-fluoro-3-methoxy-6-(trifluoromethyl)benzoate


Formula: C₁₁H₁₀F₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34530098
CAS Number: 2702116-67-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty