Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(benzyloxy)-2-methoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

52492-500MG 52492-1G 52492-5G 52492-25G
77328-710EA 228.66 USD InStock
77328-710 77352-394 77365-052 77387-140
Methyl 4-(benzyloxy)-2-methoxybenzoate ≥97%
Methyl 4-(benzyloxy)-2-methoxybenzoate
Precaución: for Research Use Only


Formula: C₁₆H₁₆O₄
MW: 272.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204271
CAS Number: 28478-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty