Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromo-4-methylanisole ≥95%

Supplier: AOB CHEM USA
Warning

52831-500MG 52831-1G 52831-5G 52831-25G
77328-810EA 135.96 USD InStock
77328-810 77344-514 77365-128 77387-214
2,5-Dibromo-4-methylanisole ≥95%
2,5-Dibromo-4-methylanisole
Precaución: for Research Use Only


Formula: C₈H₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD12547800
CAS Number: 860585-46-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty