Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-bromo-2,3-difluoro-4-methylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

53292-500MG 53292-1G 53292-5G 53292-25G
77328-972EA 278.1 USD InStock
77328-972 77352-732 77365-252 77387-312
Ethyl 5-bromo-2,3-difluoro-4-methylbenzoate ≥97%
Ethyl 5-bromo-2,3-difluoro-4-methylbenzoate


Formula: C₁₀H₉BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34530208
CAS Number: 2734775-49-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty