Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Ethoxy-2-methylnicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

53338-500MG 53338-1G 53338-5G 53338-25G
77328-986EA 238.96 USD InStock
77328-986 77352-758 77365-268 77387-322
6-Ethoxy-2-methylnicotinic acid ≥97%
6-Ethoxy-2-methylnicotinic acid
Precaución: for Research Use Only


Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21086438
CAS Number: 1355180-16-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty