Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl(3,4,5-trifluorophenyl)sulfane ≥95%

Supplier: AOB CHEM USA
Warning

53656-500MG 53656-1G 53656-5G 53656-25G
77329-076EA 250.29 USD InStock
77329-076 77352-870 77365-374 77387-388
Methyl(3,4,5-trifluorophenyl)sulfane ≥95%
Methyl(3,4,5-trifluorophenyl)sulfane


Formula: C₇H₅F₃S
Storage Temperature: Ambient
MDL Number: MFCD11617752
CAS Number: 1314966-54-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty