Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-(methylthio)-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

53836-500MG 53836-1G 53836-25G 53836-5G
77329-120EA 284.28 USD InStock
77329-120 77352-926 77387-426 77377-146
Ethyl 3-(methylthio)-5-(trifluoromethyl)benzoate ≥97%
Ethyl 3-(methylthio)-5-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₁F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD22489161
CAS Number: 2733750-52-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty