Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-(methylthio)-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOB CHEM USA
Warning

53837-500MG 53837-1G 53837-25G 53837-5G
77329-122EA 300.76 USD InStock
77329-122 77352-928 77387-428 77377-148
Methyl 3-(methylthio)-5-(trifluoromethyl)benzoate ≥97%
Methyl 3-(methylthio)-5-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₉F₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD22489113
CAS Number: 1823311-77-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty