Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethyl-5-(methylthio)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

54542-500MG 54542-1G 54542-5G 54542-25G
77329-268EA 280.16 USD InStock
77329-268 77353-088 77365-554 77387-524
3-Ethyl-5-(methylthio)benzoic acid ≥97%
3-Ethyl-5-(methylthio)benzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂S
MW: 196.266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489513
CAS Number: 1822660-94-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty