Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethyl-3,4-difluorobenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

54598-500MG 54598-1G 54598-25G 54598-5G
77329-292EA 297.67 USD InStock
77329-292 77353-134 77387-540 77377-364
2-Ethyl-3,4-difluorobenzoic acid ≥97%
2-Ethyl-3,4-difluorobenzoic acid
Precaución: for Research Use Only


Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD27938383
CAS Number: 1251164-92-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty