Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-5-fluoro-2-methylphenol ≥97%

Supplier: AOB CHEM USA
Warning

54869-500MG 54869-1G 54869-5G
77338-850EA 525.3 USD InStock
77338-850 77353-218 77377-476
4-Chloro-5-fluoro-2-methylphenol ≥97%
4-Chloro-5-fluoro-2-methylphenol
Precaución: for Research Use Only


Formula: C₇H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD23110792
CAS Number: 1395036-03-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty