Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,3-dibromo-5-methylbenzoate ≥95%

Supplier: AOB CHEM USA
Warning

54997-500MG 54997-1G 54997-5G 54997-25G
77329-370EA 121.54 USD InStock
77329-370 77343-868 77365-670 77377-500
Methyl 2,3-dibromo-5-methylbenzoate ≥95%
Methyl 2,3-dibromo-5-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₈Br₂O₂
MW: 307.967 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24107270
CAS Number: 1500168-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty