Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluoro-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOB CHEM USA
Warning

55538-500MG 55538-1G 55538-5G 55538-25G
77329-480EA 151.41 USD InStock
77329-480 77345-426 77365-776 77387-658
3-Chloro-5-fluoro-[1,1'-biphenyl]-4-carboxylic acid ≥97%
3-Chloro-5-fluoro-[1,1'-biphenyl]-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₈ClFO₂
MW: 250.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757537
CAS Number: 2901096-38-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty