Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-fluoro-2,6-dimethoxybenzoate ≥97%

Supplier: AOB CHEM USA
Warning

55586-500MG 55586-1G 55586-5G
77339-576EA 599.46 USD InStock
77339-576 77353-368 77377-642
Methyl 3-fluoro-2,6-dimethoxybenzoate ≥97%
Methyl 3-fluoro-2,6-dimethoxybenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₁FO₄
MW: 214.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28792936
CAS Number: 1803848-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty