Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-(methylthio)-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

55668-500MG 55668-1G 55668-5G 55668-25G
77329-508EA 258.53 USD InStock
77329-508 77353-392 77365-810 77387-680
2-Fluoro-3-(methylthio)-4-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-(methylthio)-4-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₆F₄O₂S
MW: 254.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31636381
CAS Number: 1262515-30-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty