Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxy-5-ethylphenol ≥95%

Supplier: AOB CHEM USA
Warning

57612-500MG 57612-1G 57612-5G
77337-952EA 454.23 USD InStock
77337-952 77353-950 77378-172
2-Ethoxy-5-ethylphenol ≥95%
2-Ethoxy-5-ethylphenol
Precaución: for Research Use Only


Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD32877187
CAS Number: 1466788-20-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty