Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-(methylsulfonyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOB CHEM USA
Warning

57681-500MG 57681-1G 57681-5G 57681-25G
77330-002EA 222.48 USD InStock
77330-002 77353-964 77366-306 77388-034
2-Fluoro-4-(methylsulfonyl)phenylboronic acid pinacol ester ≥97%
2-Fluoro-4-(methylsulfonyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₈BFO₄S
Storage Temperature: Ambient
MDL Number: MFCD18760181
CAS Number: 1384951-71-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty