Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzyloxy)-5-ethylbenzaldehyde ≥95%

Supplier: AOB CHEM USA
Warning

57751-500MG 57751-1G 57751-5G 57751-25G
77330-032EA 152.44 USD InStock
77330-032 77345-456 77366-332 77388-062
2-(Benzyloxy)-5-ethylbenzaldehyde ≥95%
2-(Benzyloxy)-5-ethylbenzaldehyde
Precaución: for Research Use Only


Formula: C₁₆H₁₆O₂
MW: 240.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31504486
CAS Number: 749934-46-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty