Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-formyl-4-isopropoxybenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

57990-500MG 57990-1G 57990-5G 57990-25G
77330-078EA 189.52 USD InStock
77330-078 77354-086 77366-372 77388-092
3-Fluoro-5-formyl-4-isopropoxybenzoic acid ≥97%
3-Fluoro-5-formyl-4-isopropoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁FO₄
MW: 226.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31422175
CAS Number: 2138215-46-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty