Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Benzyloxy)-2-bromo-4-isobutylbenzene ≥97%

Supplier: AOB CHEM USA
Warning

58111-500MG 58111-1G 58111-25G 58111-5G
77330-142EA 301.79 USD InStock
77330-142 77354-154 77388-146 77378-330
1-(Benzyloxy)-2-bromo-4-isobutylbenzene ≥97%
1-(Benzyloxy)-2-bromo-4-isobutylbenzene
Precaución: for Research Use Only


Formula: C₁₇H₁₉BrO
Storage Temperature: Ambient
MDL Number: MFCD34758457
CAS Number: 2755724-58-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty