Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxy-5-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

58342-500MG 58342-1G 58342-5G
77338-686EA 509.85 USD InStock
77338-686 77354-306 77378-420
2-Ethoxy-5-fluoro-3-methylbenzoic acid ≥97%
2-Ethoxy-5-fluoro-3-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD26734767
CAS Number: 1556338-89-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty