Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-ethoxy-2,5-dimethylbenzoate ≥97%

Supplier: AOB CHEM USA
Warning

58461-500MG 58461-1G 58461-5G
77339-612EA 603.58 USD InStock
77339-612 77354-356 77378-466
Methyl 4-ethoxy-2,5-dimethylbenzoate ≥97%
Methyl 4-ethoxy-2,5-dimethylbenzoate
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD31773130
CAS Number: 1368220-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty